missing translation for 'onlineSavingsMsg'
Learn More

(S)-5-(Pyrrolidin-2-yl)-1H-tetrazole 98.0+%, TCI America™

Catalog No. P1784100MG
Change view
Click to view available options
Quantity:
100 mg
500 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P1784100MG 100 mg
P1784500MG 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P1784100MG Supplier TCI America Supplier No. P1784100MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 33878-70-5
Molecular Formula C5H9N5
Molecular Weight (g/mol) 139.162
InChI Key XUHYQIQIENDJER-BYPYZUCNSA-N
PubChem CID 11275040
IUPAC Name 5-[(2S)-pyrrolidin-2-yl]-2H-tetrazole
SMILES C1CC(NC1)C2=NNN=N2

Specifications

Melting Point 271°C
Color White-Yellow
Quantity 100 mg
Formula Weight 139.16
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material (S)-5-(Pyrrolidin-2-yl)-1H-tetrazole