accessibility menu, dialog, popup

UserName

CAS RN 33878-70-5

IUPAC Name: 5-(pyrrolidin-2-yl)-2H-1,2,3,4-tetrazole
Synonyms: 5-(pyrrolidin-2-yl)-2H-1,2,3,4-tetrazole
Molecular Weight (g/mol): 139.16
Molecular Formula: C5H9N5
InChi Key: XUHYQIQIENDJER-UHFFFAOYNA-N
SMILES: C1CNC(C1)C1=NNN=N1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results