missing translation for 'onlineSavingsMsg'
Learn More

(S)-5,5-Diphenyl-2-methyl-3,4-propano-1,3,2-oxazaborolidine 98.0+%, TCI America™

Catalog No. D21311G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D21311G 1 g
D21315G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D21311G Supplier TCI America Supplier No. D21311G

Chemical Identifiers

CAS 112022-81-8
Molecular Formula C18H20BNO
Molecular Weight (g/mol) 277.17
MDL Number MFCD00078439
InChI Key VMKAFJQFKBASMU-KRWDZBQOSA-N
Synonym s-2-methyl-cbs-oxazaborolidine, s-methyl oxazaborolidine, s---2-methyl-cbs-oxazaborolidine, s-5,5-diphenyl-2-methyl-3,4-propano-1,3,2-oxazaborolidine, corey catalyst, s-methyl-cbs-oxazaborolidine, s-3,3-diphenyl-1-methylpyrrolidino 1,2-c-1,3,2-oxazaborole, s-me-cbs catalyst, s-methyl-cbs, s-me-cbs
PubChem CID 2734713
IUPAC Name (3aS)-1-methyl-3,3-diphenyl-hexahydropyrrolo[1,2-c][1,3,2]oxazaborole
SMILES [H][C@@]12CCCN1B(C)OC2(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Color White
Quantity 1 g
Formula Weight 277.17
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material (S)-5,5-Diphenyl-2-methyl-3,4-propano-1,3,2-oxazaborolidine