missing translation for 'onlineSavingsMsg'
Learn More

(S)-(-)-5,5',6,6',7,7',8,8'-Octahydro-1,1'-bi-2-naphthol 99.0+%, TCI America™

Catalog No. p-7134276
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
O02831G 1 g
O02835G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. O02831G Supplier TCI America Supplier No. O02831G
Only null left

Chemical Identifiers

CAS 65355-00-2
Molecular Formula C20H22O2
Molecular Weight (g/mol) 294.39
MDL Number MFCD02093485
InChI Key UTXIFKBYNJRJPH-UHFFFAOYSA-N
Synonym s---5,5',6,6',7,7',8,8'-octahydro-1,1'-bi-2-naphthol, r-+-5,5',6,6',7,7',8,8'-octahydro-1,1'-bi-2-naphthol, r-5,5',6,6',7,7',8,8'-octahydro 1,1'-binaphthalene-2,2'-diol, s-5,5',6,6',7,7',8,8'-octahydro-1,1'-binaphthalene-2,2'-diol, 5,5',6,6',7,7',8,8'-octahydro-1,1'-binaphthalene-2,2'-diol, 5,5',6,6',7,7',8,8'-octahydro-1,1'-bi-2-naphthol, 5,5',6,6',7,7',8,8'-octahydro-bi-2-naphthol, r-5,5',6,6',7,7',8,8'-octahydro-1,1'-bi-2-naphthol, s---2,2'-dihydroxy-5,5',6,6',7,7',8,8'-octahydro-1,1'-binaphthyl, 1-2-hydroxy-5,6,7,8-tetrahydronaphthalen-1-yl-5,6,7,8-tetrahydronaphthalen-2-ol
PubChem CID 3694111
IUPAC Name 5H,5'H,6H,6'H,7H,7'H,8H,8'H-[1,1'-binaphthalene]-2,2'-diol
SMILES OC1=C(C2=C(CCCC2)C=C1)C1=C(O)C=CC2=C1CCCC2

Specifications

Melting Point 167°C
Color White
Quantity 1 g
Formula Weight 294.39
Percent Purity ≥99.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material (S)-(-)-5,5′,6,6′,7,7′,8,8′-Octahydro-1,1′-bi-2-naphthol