missing translation for 'onlineSavingsMsg'
Learn More

(S)-4-tert-Butyl-2-(2-pyridyl)oxazoline 98.0+%, TCI America™

Catalog No. B41045G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B41045G 5 g
B41041G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B41045G Supplier TCI America Supplier No. B41045G
Only null left

Chemical Identifiers

CAS 117408-98-7
Molecular Formula C12H16N2O
Molecular Weight (g/mol) 204.273
MDL Number MFCD22200533
InChI Key JDOAIDZRVWJBEG-SNVBAGLBSA-N
Synonym (S)-4-(tert-Butyl)-2-(2-pyridyl)-4,5-dihydrooxazole
PubChem CID 14337774
IUPAC Name (4S)-4-tert-butyl-2-pyridin-2-yl-4,5-dihydro-1,3-oxazole
SMILES CC(C)(C)C1COC(=N1)C2=CC=CC=N2

Specifications

Melting Point 72°C
Color White
Quantity 5 g
Formula Weight 204.27
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material (S)-4-tert-Butyl-2-(2-pyridyl)oxazoline