CAS RN 117408-98-7
IUPAC Name:
2-(4-tert-butyl-4,5-dihydro-1,3-oxazol-2-yl)pyridine
Synonyms:
2-(4-tert-butyl-4,5-dihydro-1,3-oxazol-2-yl)pyridine
Molecular Weight (g/mol):
204.27
Molecular Formula:
C12H16N2O
InChi Key:
JDOAIDZRVWJBEG-UHFFFAOYNA-N
SMILES:
CC(C)(C)C1COC(=N1)C1=CC=CC=N1
1
–
1
of
1
results
1
(S)-4-tert-Butyl-2-(2-pyridyl)oxazoline 98.0+%, TCI America™
CAS: 117408-98-7 Molecular Formula: C12H16N2O Molecular Weight (g/mol): 204.273 MDL Number: MFCD22200533 InChI Key: JDOAIDZRVWJBEG-SNVBAGLBSA-N Synonym: (S)-4-(tert-Butyl)-2-(2-pyridyl)-4,5-dihydrooxazole PubChem CID: 14337774 IUPAC Name: (4S)-4-tert-butyl-2-pyridin-2-yl-4,5-dihydro-1,3-oxazole SMILES: CC(C)(C)C1COC(=N1)C2=CC=CC=N2