missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(S)-(+)-4-Phenyl-2-oxazolidinone 99.0+%, TCI America™
Supplier: TCI America P130825G
Specifications
| 131°C | |
| 99395-88-7 | |
| C9H9NO2 | |
| s-+-4-phenyl-2-oxazolidinone, s-4-phenyl-2-oxazolidinone, s-4-phenyloxazolidin-2-one, 4s-4-phenyl-1,3-oxazolidin-2-one, 2-oxazolidinone, 4-phenyl-, 4s, 4s-4-phenyloxazolidin-2-one, s-4-phenyl-2-oxazolidenone, s-ph-oxazolidinone, pubchem6076 | |
| C1C(NC(=O)O1)C2=CC=CC=C2 | |
| 25 g | |
| 730424 | |
| ≥99.0% (GC) |
| (S)-(+)-4-Phenyl-2-oxazolidinone | |
| White | |
| MFCD00043396 | |
| QDMNNMIOWVJVLY-MRVPVSSYSA-N | |
| (4S)-4-phenyl-1,3-oxazolidin-2-one | |
| 163.176 | |
| 163.18 | |
| Crystalline Powder |
Chemical Identifiers
| 99395-88-7 | |
| 163.176 | |
| QDMNNMIOWVJVLY-MRVPVSSYSA-N | |
| 730424 | |
| C1C(NC(=O)O1)C2=CC=CC=C2 |
| C9H9NO2 | |
| MFCD00043396 | |
| s-+-4-phenyl-2-oxazolidinone, s-4-phenyl-2-oxazolidinone, s-4-phenyloxazolidin-2-one, 4s-4-phenyl-1,3-oxazolidin-2-one, 2-oxazolidinone, 4-phenyl-, 4s, 4s-4-phenyloxazolidin-2-one, s-4-phenyl-2-oxazolidenone, s-ph-oxazolidinone, pubchem6076 | |
| (4S)-4-phenyl-1,3-oxazolidin-2-one |
Safety and Handling
TSCA : No