missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(S)-(+)-4-Phenyl-2-oxazolidinone 99.0+%, TCI America™
$33.56 - $52.19
Chemical Identifiers
| CAS | 99395-88-7 |
|---|---|
| Molecular Formula | C9H9NO2 |
| Molecular Weight (g/mol) | 163.176 |
| MDL Number | MFCD00043396 |
| InChI Key | QDMNNMIOWVJVLY-MRVPVSSYSA-N |
| Synonym | s-+-4-phenyl-2-oxazolidinone, s-4-phenyl-2-oxazolidinone, s-4-phenyloxazolidin-2-one, 4s-4-phenyl-1,3-oxazolidin-2-one, 2-oxazolidinone, 4-phenyl-, 4s, 4s-4-phenyloxazolidin-2-one, s-4-phenyl-2-oxazolidenone, s-ph-oxazolidinone, pubchem6076 |
| PubChem CID | 730424 |
| IUPAC Name | (4S)-4-phenyl-1,3-oxazolidin-2-one |
| SMILES | C1C(NC(=O)O1)C2=CC=CC=C2 |
Chemical Identifiers
| 99395-88-7 | |
| 163.176 | |
| QDMNNMIOWVJVLY-MRVPVSSYSA-N | |
| 730424 | |
| C1C(NC(=O)O1)C2=CC=CC=C2 |
| C9H9NO2 | |
| MFCD00043396 | |
| s-+-4-phenyl-2-oxazolidinone, s-4-phenyl-2-oxazolidinone, s-4-phenyloxazolidin-2-one, 4s-4-phenyl-1,3-oxazolidin-2-one, 2-oxazolidinone, 4-phenyl-, 4s, 4s-4-phenyloxazolidin-2-one, s-4-phenyl-2-oxazolidenone, s-ph-oxazolidinone, pubchem6076 | |
| (4S)-4-phenyl-1,3-oxazolidin-2-one |
Specifications
| 131°C | |
| White | |
| MFCD00043396 | |
| QDMNNMIOWVJVLY-MRVPVSSYSA-N | |
| (4S)-4-phenyl-1,3-oxazolidin-2-one | |
| 163.176 | |
| 163.18 | |
| Crystalline Powder |
| 99395-88-7 | |
| C9H9NO2 | |
| s-+-4-phenyl-2-oxazolidinone, s-4-phenyl-2-oxazolidinone, s-4-phenyloxazolidin-2-one, 4s-4-phenyl-1,3-oxazolidin-2-one, 2-oxazolidinone, 4-phenyl-, 4s, 4s-4-phenyloxazolidin-2-one, s-4-phenyl-2-oxazolidenone, s-ph-oxazolidinone, pubchem6076 | |
| C1C(NC(=O)O1)C2=CC=CC=C2 | |
| 5 g | |
| 730424 | |
| ≥99.0% (GC) | |
| (S)-(+)-4-Phenyl-2-oxazolidinone |
Safety and Handling
TSCA : No