missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(S)-(+)-4-Methyl-2-pentanol, 99%
Description
It is employed in the reaction of 2-methoxyethyl acetals with allylsilanes in the presence of titanium tetrachloride: regioselective carbon-oxygen bond cleavage of the unsymmetrical acetals.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
It is employed in the reaction of 2-methoxyethyl acetals with allylsilanes in the presence of titanium tetrachloride: regioselective carbon-oxygen bond cleavage of the unsymmetrical acetals.
Solubility
Slightly soluble in water. Soluble in alcohol.
Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Keep ignition sources away. Protect from electrostatic charges.
It is employed in the reaction of 2-methoxyethyl acetals with allylsilanes in the presence of titanium tetrachloride: regioselective carbon-oxygen bond cleavage of the unsymmetrical acetals.
Solubility
Slightly soluble in water. Soluble in alcohol.
Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Keep ignition sources away. Protect from electrostatic charges.
Specifications
Specifications
| Density | 0.802 |
| Boiling Point | 130°C to 131°C |
| Flash Point | 41°C (105°F) |
| Odor | Slightly Sweetish |
| Refractive Index | 1.411 |
| Quantity | 250 mg |
| UN Number | UN2053 |
| Beilstein | 1718991 |
| Synonym | s-+-4-methyl-2-pentanol, 2s-4-methylpentan-2-ol, s-+-4-methylpentan-2-ol, 4-methyl s-2-pentanol, s-4-methyl-pentan-2-ol, 2s-4-methyl-2-pentanol, s-1,3-dimethyl-1-butanol, 2-pentanol,4-methyl-, 2s, 2-pentanol, 4-methyl-, 2s, unii-8u34xjk0r0 component |
| Solubility Information | Slightly soluble in water. Soluble in alcohol. |
| Show More |
Safety and Handling
| Hazard Category | H226-H335 |
| Hazard Statement | GHS H Statement H226-H335 Flammable liquid and vapor. May cause respiratory irritation. |
| Precautionary Statement | P210-P233-P235-P240-P241-P242-P243-P261-P271-P280-P303+P361+P353-P304+P340-P312-P370+P378q-P501c |
| DOTInformation | Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: METHYL ISOBUTYL CARBINOL |
| TSCA | No |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only