accessibility menu, dialog, popup

UserName

CH3H3CCH3HO

CAS RN 14898-80-7

IUPAC Name: 4-methylpentan-2-ol
Synonyms: 4-methyl-2-pentanol
Molecular Weight (g/mol): 102.18
Molecular Formula: C6H14O
InChi Key: WVYWICLMDOOCFB-UHFFFAOYNA-N
SMILES: CC(C)CC(C)O
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (1)
  • (1)