missing translation for 'onlineSavingsMsg'
Learn More

(S)-(-)-4-(Methoxymethyl)-1,3-dioxolan-2-one 98.0+%, TCI America™

Catalog No. p-7132746
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M14565G 5 g
M145625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M14565G Supplier TCI America Supplier No. M14565G

Chemical Identifiers

CAS 135682-18-7
Molecular Formula C5H8O4
Molecular Weight (g/mol) 132.12
MDL Number MFCD01321302
InChI Key DNSGQMOSYDHNHO-UHFFFAOYNA-N
Synonym (S)-(-)-4-(Methoxymethyl)-2-oxo-1,3-dioxolane, (S)-(-)-3-Methoxypropylene Carbonate
PubChem CID 11007941
IUPAC Name 4-(methoxymethyl)-1,3-dioxolan-2-one
SMILES COCC1COC(=O)O1

Specifications

Color Yellow
Boiling Point 106°C
Quantity 5 g
Formula Weight 132.12
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material (S)-(-)-4-(Methoxymethyl)-1,3-dioxolan-2-one