accessibility menu, dialog, popup

UserName

CH3OOOO

CAS RN 135682-18-7

IUPAC Name: 4-(methoxymethyl)-1,3-dioxolan-2-one
Synonyms: 4-(methoxymethyl)-1,3-dioxolan-2-one
Molecular Weight (g/mol): 132.12
Molecular Formula: C5H8O4
InChi Key: DNSGQMOSYDHNHO-UHFFFAOYNA-N
SMILES: COCC1COC(=O)O1
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results