missing translation for 'onlineSavingsMsg'
Learn More

(S)-4-Isopropyl-2-oxazolidinone 98.0+%, TCI America™

Supplier:  TCI America I045125G

Quantity 25 g

 View more versions of this product

Catalog No. I045125G


Only null left

Description

Description

Specifications

Chemical Identifiers

17016-83-0
129.159
YBUPWRYTXGAWJX-RXMQYKEDSA-N
7157133
CC(C)C1COC(=O)N1
C6H11NO2
MFCD00010847
s-4-isopropyl-2-oxazolidinone, s---4-isopropyl-2-oxazolidinone, 4s---4-isopropyl-2-oxazolidinone, s-4-isopropyloxazolidin-2-one, 4s-4-isopropyl-1,3-oxazolidin-2-one, 2-oxazolidinone, 4-1-methylethyl-, 4s, 4s-4-propan-2-yl-1,3-oxazolidin-2-one, 4s---isopropyl-2-oxazolidinone, 4 s-1-methylethyl-2-oxazolidinone, 4s---4-isopropyl-2-oxazolidinon
(4S)-4-propan-2-yl-1,3-oxazolidin-2-one

Specifications

72°C
17016-83-0
C6H11NO2
s-4-isopropyl-2-oxazolidinone, s---4-isopropyl-2-oxazolidinone, 4s---4-isopropyl-2-oxazolidinone, s-4-isopropyloxazolidin-2-one, 4s-4-isopropyl-1,3-oxazolidin-2-one, 2-oxazolidinone, 4-1-methylethyl-, 4s, 4s-4-propan-2-yl-1,3-oxazolidin-2-one, 4s---isopropyl-2-oxazolidinone, 4 s-1-methylethyl-2-oxazolidinone, 4s---4-isopropyl-2-oxazolidinon
CC(C)C1COC(=O)N1
25 g
7157133
≥98.0% (GC)
(S)-4-Isopropyl-2-oxazolidinone
White-Yellow
MFCD00010847
YBUPWRYTXGAWJX-RXMQYKEDSA-N
(4S)-4-propan-2-yl-1,3-oxazolidin-2-one
129.159
129.16
Crystalline Powder
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

TSCA : No

SDS
Documents

Documents

Promotions

Promotions