accessibility menu, dialog, popup

UserName

CH3H3C(S)OOHN

CAS RN 17016-83-0

IUPAC Name: (4S)-4-(propan-2-yl)-1,3-oxazolidin-2-one
Synonyms: (4S)-4-isopropyl-1,3-oxazolidin-2-one
Molecular Weight (g/mol): 129.16
Molecular Formula: C6H11NO2
InChi Key: YBUPWRYTXGAWJX-RXMQYKEDSA-N
SMILES: CC(C)[C@H]1COC(=O)N1
Molecular Weight (g/mol) 
  • (3)
  • (3)
Percent Purity 
  • (3)
  • (3)
Quantity 
  • (2)
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (3)
  • (3)
  • (3)
  • (3)
  • (3)
  • (2)
  • (2)
  • (2)