missing translation for 'onlineSavingsMsg'
Learn More

(S)-4-Isopropyl-2-oxazolidinone 98.0+%, TCI America™

Catalog No. I04511G
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I04511G 1 g
I04515G 5 g
I045125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. I04511G Supplier TCI America Supplier No. I04511G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 17016-83-0
Molecular Formula C6H11NO2
Molecular Weight (g/mol) 129.159
MDL Number MFCD00010847
InChI Key YBUPWRYTXGAWJX-RXMQYKEDSA-N
Synonym s-4-isopropyl-2-oxazolidinone, s---4-isopropyl-2-oxazolidinone, 4s---4-isopropyl-2-oxazolidinone, s-4-isopropyloxazolidin-2-one, 4s-4-isopropyl-1,3-oxazolidin-2-one, 2-oxazolidinone, 4-1-methylethyl-, 4s, 4s-4-propan-2-yl-1,3-oxazolidin-2-one, 4s---isopropyl-2-oxazolidinone, 4 s-1-methylethyl-2-oxazolidinone, 4s---4-isopropyl-2-oxazolidinon
PubChem CID 7157133
IUPAC Name (4S)-4-propan-2-yl-1,3-oxazolidin-2-one
SMILES CC(C)C1COC(=O)N1

Specifications

Melting Point 72°C
Color White-Yellow
Quantity 1 g
Formula Weight 129.16
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material (S)-4-Isopropyl-2-oxazolidinone