missing translation for 'onlineSavingsMsg'
Learn More

(S)-4-Fluorophenylglycine, 95%, (98% ee), Thermo Scientific Chemicals

Catalog No. AC362050010
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC362050010 1 g Glass bottle
AC362050050 5 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC362050010 Supplier Thermo Scientific Chemicals Supplier No. 362050010

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 19883-57-9 | Molecular Formula: C8H8FNO2 | Molecular Weight: 169.16 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 19883-57-9
Molecular Formula C8H8FNO2
Molecular Weight (g/mol) 169.16
MDL Number MFCD00213799
InChI Key JKFYKCYQEWQPTM-ZETCQYMHSA-N
Synonym s-4-fluorophenylglycine, s-2-amino-2-4-fluorophenyl acetic acid, 4-fluoro-l-alpha-phenylglycine, l-4-fluorophenylglycine, 4-fluoro-l-phenylglycine, l +-4-fluorophenylglycine, 2s-2-amino-2-4-fluorophenyl acetic acid, s-+-4-fluorophenyl glycine, p-fluorophenyl glycine, 4-fluorophenylglycine
PubChem CID 853015
IUPAC Name (2S)-2-amino-2-(4-fluorophenyl)acetic acid
SMILES [NH3+][C@H](C([O-])=O)C1=CC=C(F)C=C1

Specifications

Melting Point >300°C
Color White
Enantiomeric Excess 97.5% min. (HPLC)
Infrared Spectrum Authentic
Packaging Glass bottle
Quantity 1 g
Formula Weight 169.15
Percent Purity 95%,(98% ee)
Physical Form Fluffy Powder
Chemical Name or Material (S)-4-Fluorophenylglycine, 95%, (98% e.e.)
Hazard Category HYGROSCOPIC
TSCA TSCA

RUO – Research Use Only