accessibility menu, dialog, popup

UserName

CAS RN 19883-57-9

IUPAC Name: (2S)-2-azaniumyl-2-(4-fluorophenyl)acetate
Synonyms: (2S)-2-ammonio-2-(4-fluorophenyl)acetate
Molecular Weight (g/mol): 169.16
Molecular Formula: C8H8FNO2
InChi Key: JKFYKCYQEWQPTM-ZETCQYMHSA-N
SMILES: [NH3+][C@H](C([O-])=O)C1=CC=C(F)C=C1
Percent Purity 
  • (2)
  • (2)
Physical Form 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)