missing translation for 'onlineSavingsMsg'
Learn More

(S)-4-Cyano-4'-(2-methylbutyl)biphenyl 98.0+%, TCI America™

Catalog No. C29131G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C29131G 1 g
C29135G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C29131G Supplier TCI America Supplier No. C29131G
Only null left

Chemical Identifiers

CAS 63799-11-1
Molecular Formula C18H19N
Molecular Weight (g/mol) 249.357
MDL Number MFCD00192280
InChI Key DNJQGRFZQMOYGM-AWEZNQCLSA-N
Synonym s-4'-2-methylbutyl-1,1'-biphenyl-4-carbonitrile, s-4-cyano-4'-2-methylbutyl biphenyl, 4-s-2-methylbutyl-4'-cyanobiphenyl, s-4'-2-methylbutyl 1,1'-biphenyl-4-carbonitrile, 4-4-2s-2-methylbutyl phenyl benzonitrile, 1,1'-biphenyl-4-carbonitrile,4'-2s-2-methylbutyl, 1,1'-biphenyl-4-carbonitrile, 4'-2s-2-methylbutyl, 4-cyano-4'-2-methylbut-1-yl biphenyl
PubChem CID 114240
IUPAC Name 4-[4-[(2S)-2-methylbutyl]phenyl]benzonitrile
SMILES CCC(C)CC1=CC=C(C=C1)C2=CC=C(C=C2)C#N

Specifications

Color Yellow
Quantity 1 g
Formula Weight 249.36
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material (S)-4-Cyano-4′-(2-methylbutyl)biphenyl