missing translation for 'onlineSavingsMsg'
Learn More

(S)-4-(Boc-amino)-2-(Fmoc-amino)butyric acid, 96%

CAS: 125238-99-5 | C24H28N2O6 | 440.50 g/mol

Supplier:  Thermo Scientific Chemicals H5199006

 View more versions of this product

Catalog No. AAH5199006


Only null left
Explore available promotions

Description

Description

It finds its application in combination with the Nγ-Z derivative for the synthesis of polymyxin analogs.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It finds its application in combination with the Nγ-Z derivative for the synthesis of polymyxin analogs.

Solubility
Slightly soluble in water.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Keep it refrigerated.
Specifications

Specifications

(S)-4-(Boc-amino)-2-(Fmoc-amino)butyric acid
111°C to 113°C
MFCD00151941
6250328
Slightly soluble in water.
−12° (c=1 in Methanol)
(2S)-2-(9H-fluoren-9-ylmethoxycarbonylamino)-4-[(2-methylpropan-2-yl)oxycarbonylamino]butanoic acid
2756101
96%
125238-99-5
C24H28N2O6
5 g
fmoc-dab boc-oh, s-2-9h-fluoren-9-yl methoxy carbonyl amino-4-tert-butoxycarbonyl amino butanoic acid, fmoc-n4-boc-l-2,4-diaminobutyric acid, 2s-4-tert-butoxycarbonyl amino-2-9h-fluoren-9-ylmethoxy carbonyl amino butanoic acid, fmoc-l-dab boc-oh, fmoc-dab boc, ambotzfaa1364, n-fmoc-n'-boc-l-2,4-diaminobutyric acid, na-fmoc-n-boc-l-2,4-diaminobutyric acid, fmoc-dab boc-oh hplc
LIWKOFAHRLBNMG-UHFFFAOYNA-N
CC(C)(C)OC(=O)NCCC(NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12)C(O)=O
440.50
440.5

Chemical Identifiers

125238-99-5
440.50
LIWKOFAHRLBNMG-UHFFFAOYNA-N
2756101
CC(C)(C)OC(=O)NCCC(NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12)C(O)=O
C24H28N2O6
MFCD00151941
fmoc-dab boc-oh, s-2-9h-fluoren-9-yl methoxy carbonyl amino-4-tert-butoxycarbonyl amino butanoic acid, fmoc-n4-boc-l-2,4-diaminobutyric acid, 2s-4-tert-butoxycarbonyl amino-2-9h-fluoren-9-ylmethoxy carbonyl amino butanoic acid, fmoc-l-dab boc-oh, fmoc-dab boc, ambotzfaa1364, n-fmoc-n'-boc-l-2,4-diaminobutyric acid, na-fmoc-n-boc-l-2,4-diaminobutyric acid, fmoc-dab boc-oh hplc
(2S)-2-(9H-fluoren-9-ylmethoxycarbonylamino)-4-[(2-methylpropan-2-yl)oxycarbonylamino]butanoic acid
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only