missing translation for 'onlineSavingsMsg'
Learn More

(S)-4-(Boc-amino)-2-(Fmoc-amino)butyric acid, 96%

Catalog No. AAH51990MD
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH51990MD 250 mg
AAH5199003 1 g
AAH5199006 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAH51990MD Supplier Thermo Scientific Chemicals Supplier No. H51990MD

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 125238-99-5 | C24H28N2O6 | 440.50 g/mol

It finds its application in combination with the Nγ-Z derivative for the synthesis of polymyxin analogs.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It finds its application in combination with the Nγ-Z derivative for the synthesis of polymyxin analogs.

Solubility
Slightly soluble in water.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Keep it refrigerated.

Chemical Identifiers

CAS 125238-99-5
Molecular Formula C24H28N2O6
Molecular Weight (g/mol) 440.50
MDL Number MFCD00151941
InChI Key LIWKOFAHRLBNMG-UHFFFAOYNA-N
Synonym fmoc-dab boc-oh, s-2-9h-fluoren-9-yl methoxy carbonyl amino-4-tert-butoxycarbonyl amino butanoic acid, fmoc-n4-boc-l-2,4-diaminobutyric acid, 2s-4-tert-butoxycarbonyl amino-2-9h-fluoren-9-ylmethoxy carbonyl amino butanoic acid, fmoc-l-dab boc-oh, fmoc-dab boc, ambotzfaa1364, n-fmoc-n'-boc-l-2,4-diaminobutyric acid, na-fmoc-n-boc-l-2,4-diaminobutyric acid, fmoc-dab boc-oh hplc
PubChem CID 2756101
IUPAC Name (2S)-2-(9H-fluoren-9-ylmethoxycarbonylamino)-4-[(2-methylpropan-2-yl)oxycarbonylamino]butanoic acid
SMILES CC(C)(C)OC(=O)NCCC(NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12)C(O)=O

Specifications

Melting Point 111°C to 113°C
Quantity 250 mg
Beilstein 6250328
Solubility Information Slightly soluble in water.
Optical Rotation −12° (c=1 in Methanol)
Formula Weight 440.5
Percent Purity 96%
Chemical Name or Material (S)-4-(Boc-amino)-2-(Fmoc-amino)butyric acid
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only