missing translation for 'onlineSavingsMsg'
Learn More

(S)-4-Benzyloxycarbonylamino-2-(Fmoc-amino)butyric acid, 95%

Catalog No. AAH6372606
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6372606 5 g
AAH6372603 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6372606 Supplier Thermo Scientific Chemicals Supplier No. H6372606
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 252049-08-4 | C27H28N2O5 | 460.53 g/mol

(S)-4-Benzyloxycarbonylamino-2-(Fmoc-amino)butyric acid a substituted fluorene compound for proteomics research. Also used in agrochemical, pharmaceutical and dyestuff field.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
(S)-4-Benzyloxycarbonylamino-2-(Fmoc-amino)butyric acid a substituted fluorene compound for proteomics research. Also used in agrochemical, pharmaceutical and dyestuff field.

Solubility
Slightly soluble in water.

Notes
Store in freezer at(-20°C). Keep container tightly closed. Store away from oxidizing agent.

Chemical Identifiers

CAS 252049-08-4
Molecular Formula C27H28N2O5
Molecular Weight (g/mol) 460.53
MDL Number MFCD02259487
InChI Key KKUPBFMKUPMFRE-VWLOTQADSA-N
Synonym s-2-9h-fluoren-9-yl methoxy carbonyl amino-4-benzyloxy methyl amino butanoic acid, 2s-4-benzyloxy methyl amino-2-9h-fluoren-9-yl methoxy carbonyl amino butanoic acid, n-a-fmoc-n-?-z-l-2,4-diaminobutyric acid, 2s-4-benzyloxy methyl amino-2-9h-fluoren-9-ylmethoxy carbonyl amino butanoic acid
PubChem CID 70700347
IUPAC Name (2S)-2-(9H-fluoren-9-ylmethoxycarbonylamino)-4-(phenylmethoxymethylamino)butanoic acid
SMILES C1=CC=C(C=C1)COCNCCC(C(=O)O)NC(=O)OCC2C3=CC=CC=C3C4=CC=CC=C24

Specifications

Color White
Quantity 5 g
Solubility Information Slightly soluble in water.
Formula Weight 474.51
Percent Purity 95%
Physical Form Powder
Chemical Name or Material (S)-4-Benzyloxycarbonylamino-2-(Fmoc-amino)butyric acid
TSCA No
storageNote1 -30° to -10°C
Recommended Storage Store at -20°C

RUO – Research Use Only