missing translation for 'onlineSavingsMsg'
Learn More

(S)-(+)-4-Benzyl-3-propionyl-2-oxazolidinone 96.0+%, TCI America™

Catalog No. b21665g
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B21665G 5 g
B21661G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B21665G Supplier TCI America Supplier No. B21665G

Chemical Identifiers

CAS 101711-78-8
Molecular Formula C13H15NO3
Molecular Weight (g/mol) 233.27
MDL Number MFCD00269688
InChI Key WHOBYFHKONUTMW-NSHDSACASA-N
Synonym s-4-benzyl-3-propionyloxazolidin-2-one, s-+-4-benzyl-3-propionyl-2-oxazolidinone, s-4-benzyl-3-propionyl-2-oxazolidinone, 4s-+-4-benzyl-3-propionyl-2-oxazolidinone, 4s-4-benzyl-3-propanoyl-1,3-oxazolidin-2-one, n-propionyl-4s-benzyl-2-oxazolidinone, s---4-benzyl-3-propionyl-2-oxazolidinone, 2-oxazolidinone, 3-1-oxopropyl-4-phenylmethyl-, 4s, pubchem8129, ksc498c8l
PubChem CID 2733696
IUPAC Name (4S)-4-benzyl-3-propanoyl-1,3-oxazolidin-2-one
SMILES CCC(=O)N1[C@@H](CC2=CC=CC=C2)COC1=O

Specifications

Melting Point 45°C
Color White-Yellow
Quantity 5 g
Formula Weight 233.27
Percent Purity ≥96.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material (S)-(+)-4-Benzyl-3-propionyl-2-oxazolidinone