missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(S)-4-Benzyl-2-oxazolidinone, 99%, >99% ee
CAS: 90719-32-7 | C10H11NO2 | 177.20 g/mol
$142.08
Chemical Identifiers
| CAS | 90719-32-7 |
|---|---|
| Molecular Formula | C10H11NO2 |
| Molecular Weight (g/mol) | 177.20 |
| MDL Number | MFCD00064496 |
| InChI Key | OJOFMLDBXPDXLQ-VIFPVBQESA-N |
| Synonym | s-4-benzyl-2-oxazolidinone, s-4-benzyloxazolidin-2-one, 4s-4-benzyl-1,3-oxazolidin-2-one, s---4-benzyl-2-oxazolidinone, s-4-benzyl-oxazolidin-2-one, 2-oxazolidinone, 4-phenylmethyl-, 4s, s-+-4-benzyl-2-oxazolidinone, s-4-phenylmethyl-2-oxazolidinone, 4s-4-phenylmethyl-1,3-oxazolidin-2-one |
| PubChem CID | 736225 |
| IUPAC Name | (4S)-4-benzyl-1,3-oxazolidin-2-one |
| SMILES | O=C1N[C@@H](CC2=CC=CC=C2)CO1 |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity & Availability | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity & Availability | ||||
|
AC402160025
|
N/A | 2.5 g | Glass Bottle |
Each for $142.08
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 90719-32-7 | |
| 177.20 | |
| OJOFMLDBXPDXLQ-VIFPVBQESA-N | |
| 736225 | |
| O=C1N[C@@H](CC2=CC=CC=C2)CO1 |
| C10H11NO2 | |
| MFCD00064496 | |
| s-4-benzyl-2-oxazolidinone, s-4-benzyloxazolidin-2-one, 4s-4-benzyl-1,3-oxazolidin-2-one, s---4-benzyl-2-oxazolidinone, s-4-benzyl-oxazolidin-2-one, 2-oxazolidinone, 4-phenylmethyl-, 4s, s-+-4-benzyl-2-oxazolidinone, s-4-phenylmethyl-2-oxazolidinone, 4s-4-phenylmethyl-1,3-oxazolidin-2-one | |
| (4S)-4-benzyl-1,3-oxazolidin-2-one |
Specifications
| 87.0°C to 90.0°C | |
| White to Yellow | |
| 98.5% min. (HPLC) | |
| MFCD00064496 | |
| s-4-benzyl-2-oxazolidinone, s-4-benzyloxazolidin-2-one, 4s-4-benzyl-1,3-oxazolidin-2-one, s---4-benzyl-2-oxazolidinone, s-4-benzyl-oxazolidin-2-one, 2-oxazolidinone, 4-phenylmethyl-, 4s, s-+-4-benzyl-2-oxazolidinone, s-4-phenylmethyl-2-oxazolidinone, 4s-4-phenylmethyl-1,3-oxazolidin-2-one | |
| OJOFMLDBXPDXLQ-VIFPVBQESA-N | |
| − 62.00 (20.00°C C=1,CHCl3) | |
| 177.20 | |
| 177.2 | |
| Crystalline Powder |
| 90719-32-7 | |
| Authentic | |
| C10H11NO2 | |
| − 62.00 | |
| Solubility in water: insoluble | |
| O=C1N[C@@H](CC2=CC=CC=C2)CO1 | |
| (4S)-4-benzyl-1,3-oxazolidin-2-one | |
| 736225 | |
| 99%,>99% ee | |
| (S)-4-Benzyl-2-oxazolidinone |
Safety and Handling
GHS Signal Word: Warning
RUO – Research Use Only