missing translation for 'onlineSavingsMsg'
Learn More

(S)-4-Benzyl-2-oxazolidinone, 99%, >99% ee

Catalog No. AC402160025
Change view
Click to view available options
Quantity:
2.5 g
25 g
Packaging:
Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC402160025 2.5 g Glass Bottle
AC402160250 25 g Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC402160025 Supplier Thermo Scientific Chemicals Supplier No. 402160025

CAS: 90719-32-7 | C10H11NO2 | 177.20 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 90719-32-7
Molecular Formula C10H11NO2
Molecular Weight (g/mol) 177.20
MDL Number MFCD00064496
InChI Key OJOFMLDBXPDXLQ-VIFPVBQESA-N
Synonym s-4-benzyl-2-oxazolidinone, s-4-benzyloxazolidin-2-one, 4s-4-benzyl-1,3-oxazolidin-2-one, s---4-benzyl-2-oxazolidinone, s-4-benzyl-oxazolidin-2-one, 2-oxazolidinone, 4-phenylmethyl-, 4s, s-+-4-benzyl-2-oxazolidinone, s-4-phenylmethyl-2-oxazolidinone, 4s-4-phenylmethyl-1,3-oxazolidin-2-one
PubChem CID 736225
IUPAC Name (4S)-4-benzyl-1,3-oxazolidin-2-one
SMILES O=C1N[C@@H](CC2=CC=CC=C2)CO1

Specifications

Melting Point 87.0°C to 90.0°C
Color White to Yellow
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (HPLC)
Packaging Glass Bottle
Specific Rotation − 62.00
Solubility Information Solubility in water: insoluble
Specific Rotation Condition − 62.00 (20.00°C C=1,CHCl3)
Quantity 2.5 g
Formula Weight 177.2
Percent Purity 99%,>99% ee
Physical Form Crystalline Powder
Chemical Name or Material (S)-4-Benzyl-2-oxazolidinone
Show More Show Less
Hazard Category GHS Signal Word: Warning

RUO – Research Use Only