missing translation for 'onlineSavingsMsg'
Learn More

(S)-4-Amino-2-(Boc-amino)butyric acid, 97%

CAS: 25691-37-6 | C9H18N2O4 | 218.253 g/mol

$91.96 - $349.53

Chemical Identifiers

CAS 25691-37-6
Molecular Formula C9H18N2O4
Molecular Weight (g/mol) 218.253
MDL Number MFCD00236841
InChI Key MDCPCLPRWLKUIQ-LURJTMIESA-N
Synonym boc-l-2,4-diaminobutyric acid, boc-dab-oh, s-4-amino-2-tert-butoxycarbonyl amino butanoic acid, s-4-amino-2-tert-butoxycarbonylamino butanoic acid, n2-boc-s-2,4-diamino-butanoic acid, s-4-amino-2-boc-amino butyric acid, 2s-4-amino-2-tert-butoxycarbonyl amino butanoic acid, n-boc-s-2,4-diamino-butanoic acid, butanoic acid, 4-amino-2-1,1-dimethylethoxy carbonyl amino-, 2s, s-4-amino-2-tert-butoxycarbonylamino butyric acid
PubChem CID 7019666
IUPAC Name (2S)-4-amino-2-[(2-methylpropan-2-yl)oxycarbonylamino]butanoic acid
SMILES CC(C)(C)OC(=O)NC(CCN)C(=O)O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH6245003
View Documents
Thermo Scientific Chemicals
H6245003
1 g
Each for $91.96
Only null left
 
AAH6245006
View Documents
Thermo Scientific Chemicals
H6245006
5 g
Each for $349.53
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

25691-37-6
218.253
MDCPCLPRWLKUIQ-LURJTMIESA-N
7019666
CC(C)(C)OC(=O)NC(CCN)C(=O)O
C9H18N2O4
MFCD00236841
boc-l-2,4-diaminobutyric acid, boc-dab-oh, s-4-amino-2-tert-butoxycarbonyl amino butanoic acid, s-4-amino-2-tert-butoxycarbonylamino butanoic acid, n2-boc-s-2,4-diamino-butanoic acid, s-4-amino-2-boc-amino butyric acid, 2s-4-amino-2-tert-butoxycarbonyl amino butanoic acid, n-boc-s-2,4-diamino-butanoic acid, butanoic acid, 4-amino-2-1,1-dimethylethoxy carbonyl amino-, 2s, s-4-amino-2-tert-butoxycarbonylamino butyric acid
(2S)-4-amino-2-[(2-methylpropan-2-yl)oxycarbonylamino]butanoic acid

Specifications

25691-37-6
C9H18N2O4
1 g
boc-l-2,4-diaminobutyric acid, boc-dab-oh, s-4-amino-2-tert-butoxycarbonyl amino butanoic acid, s-4-amino-2-tert-butoxycarbonylamino butanoic acid, n2-boc-s-2,4-diamino-butanoic acid, s-4-amino-2-boc-amino butyric acid, 2s-4-amino-2-tert-butoxycarbonyl amino butanoic acid, n-boc-s-2,4-diamino-butanoic acid, butanoic acid, 4-amino-2-1,1-dimethylethoxy carbonyl amino-, 2s, s-4-amino-2-tert-butoxycarbonylamino butyric acid
CC(C)(C)OC(=O)NC(CCN)C(=O)O
218.253
218.25
Powder
White
MFCD00236841
4745487
MDCPCLPRWLKUIQ-LURJTMIESA-N
(2S)-4-amino-2-[(2-methylpropan-2-yl)oxycarbonylamino]butanoic acid
7019666
97%
(S)-4-Amino-2-(Boc-amino)butyric acid
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H318-H315
Causes serious eye damage.
Causes skin irritation.

P264b-P280-P302+P352-P305+P351+P338-P310-P332+P313-P362

H315-H318

TSCA : No

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only