missing translation for 'onlineSavingsMsg'
Learn More

S-[4-[(4-Nitrobenzyl)oxy]phenethyl]isothiourea Methanesulfonate 98.0+%, TCI America™

Catalog No. N096650MG
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N096650MG 50 mg
N096610MG 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. N096650MG Supplier TCI America Supplier No. N096650MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 182004-65-5
Molecular Formula C17H21N3O6S2
Molecular Weight (g/mol) 427.49
MDL Number MFCD00952138
InChI Key WGIKEBHIKKWJLG-UHFFFAOYSA-N
Synonym kb-r7943 mesylate, 2-2-4-4-nitrobenzyloxy phenyl ethyl isothiourea mesylate, 4-4-nitrobenzyl oxy phenethyl carbamimidothioate methanesulfonate, s-4-4-nitrobenzyl oxy phenethyl isothiourea methanesulfonate, 2-2-4-4-nitrobenzyloxy phenyl ethyl isothiourea, methane sulfonate, 2-4-4-nitrophenyl methoxy phenyl ethyl sulfanylmethanimidamide; methanesulfonic acid, d0f4fk
PubChem CID 9823846
IUPAC Name methanesulfonic acid;2-[4-[(4-nitrophenyl)methoxy]phenyl]ethyl carbamimidothioate
SMILES CS(=O)(=O)O.C1=CC(=CC=C1CCSC(=N)N)OCC2=CC=C(C=C2)[N+](=O)[O-]

Specifications

Color Yellow
Quantity 50 mg
Formula Weight 427.49
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material S-[4-[(4-Nitrobenzyl)oxy]phenethyl]isothiourea Methanesulfonate
TSCA No
Recommended Storage Refrigerator