missing translation for 'onlineSavingsMsg'
Learn More

(S)-4-(2-Hydroxyethyl)-2,2-diisopropyl-1,3-dioxolane 95.0+%, TCI America™

Catalog No. D40771G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D40771G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D40771G Supplier TCI America Supplier No. D40771G
Only null left

Chemical Identifiers

CAS 151223-12-0
Molecular Formula C11H22O3
Molecular Weight (g/mol) 202.294
InChI Key XFPZNMFFBDUEGH-JTQLQIEISA-N
Synonym (S)-2-(2,2-Diisopropyl-1,3-dioxolan-4-yl)ethanol
PubChem CID 54467051
IUPAC Name 2-[(4S)-2,2-di(propan-2-yl)-1,3-dioxolan-4-yl]ethanol
SMILES CC(C)C1(OCC(O1)CCO)C(C)C

Specifications

Color Colorless
Quantity 1 g
Formula Weight 202.29
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material (S)-4-(2-Hydroxyethyl)-2,2-diisopropyl-1,3-dioxolane