CAS RN 151223-12-0
IUPAC Name:
2-[2,2-bis(propan-2-yl)-1,3-dioxolan-4-yl]ethan-1-ol
Synonyms:
2-(2,2-diisopropyl-1,3-dioxolan-4-yl)ethanol
Molecular Weight (g/mol):
202.29
Molecular Formula:
C11H22O3
InChi Key:
XFPZNMFFBDUEGH-UHFFFAOYNA-N
SMILES:
CC(C)C1(OCC(CCO)O1)C(C)C
1
–
1
of
1
results
1
(S)-4-(2-Hydroxyethyl)-2,2-diisopropyl-1,3-dioxolane 95.0+%, TCI America™
CAS: 151223-12-0 Molecular Formula: C11H22O3 Molecular Weight (g/mol): 202.294 InChI Key: XFPZNMFFBDUEGH-JTQLQIEISA-N Synonym: (S)-2-(2,2-Diisopropyl-1,3-dioxolan-4-yl)ethanol PubChem CID: 54467051 IUPAC Name: 2-[(4S)-2,2-di(propan-2-yl)-1,3-dioxolan-4-yl]ethanol SMILES: CC(C)C1(OCC(O1)CCO)C(C)C