missing translation for 'onlineSavingsMsg'
Learn More

(S)-4-(1-Aminoethyl)phenol, 97%

Catalog No. p-7054846
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6326903 1 g
AAH6326906 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6326903 Supplier Thermo Scientific Chemicals Supplier No. H6326903

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 221670-72-0 | C8H11NO | 137.18 g/mol

(S)-4-(1-Aminoethyl)phenol is used as an intermediate in organic synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
(S)-4-(1-Aminoethyl)phenol is used as an intermediate in organic synthesis.

Notes
Incompatible with oxidizing agents.

Chemical Identifiers

CAS 221670-72-0
Molecular Formula C8H11NO
Molecular Weight (g/mol) 137.18
MDL Number MFCD03844646
InChI Key CDQPLIAKRDYOCB-UHFFFAOYNA-N
PubChem CID 41097924
IUPAC Name 4-[(1S)-1-aminoethyl]phenol
SMILES CC(N)C1=CC=C(O)C=C1

Specifications

Color White
Quantity 1 g
UN Number UN3259
Synonym 4-1s-1-aminoethyl phenol, s-4-1-aminoethyl phenol, phenol, 4-1s-1-aminoethyl, s-4-1-aminoethyl-phenol, s-4-1-amino-ethyl-phenol, s-4-1-amino-aethyl-phenol, phenol,4-1s-1-aminoethyl, s-4-hydroxy-alpha-methylbenzylamine, phenol, 4-1s-1-aminoethyl-9ci, s-4-hydroxy-? inverted exclamation marka-methylbenzylamine
Formula Weight 137.18
Percent Purity 97%
Physical Form Solid
Chemical Name or Material (S)-4-(1-Aminoethyl)phenol
Hazard Category H302-H314-H335
Hazard Statement GHS H Statement
H301-H314-H318
Toxic if swallowed.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c
DOTInformation Transport Hazard Class: 8; Packaging Group: III; Proper Shipping Name: AMINES, SOLID, CORROSIVE, N.O.S.
TSCA No
storageNote1 2° to 8°C
Recommended Storage Keep cold

RUO – Research Use Only