missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(S)-(-)-3-tert-Butoxycarbonyl-4-methoxycarbonyl-2,2-dimethyl-1,3-oxazolidine 96.0+%, TCI America™
Supplier: TCI America B17555G
Specifications
| (S)-(-)-3-tert-Butoxycarbonyl-4-methoxycarbonyl-2,2-dimethyl-1,3-oxazolidine | |
| Yellow | |
| MFCD00192279 | |
| s---3-tert-butoxycarbonyl-4-methoxycarbonyl-2,2-dimethyl-1,3-oxazolidine, 3-tert-butyl 4-methyl 4s-2,2-dimethyl-1,3-oxazolidine-3,4-dicarboxylate, methyl s---3-boc-2,2-dimethyl-4-oxazolidinecarboxylate, s-3-boc-2,2-dimethyl-oxazolidine-4-carboxylic acid methyl ester, s-3-tert-butyl 4-methyl 2,2-dimethyloxazolidine-3,4-dicarboxylate, methyl-s-3-tert-butoxycarbonyl-2,2-dimethyl-4-oxazolidinecarboxylate, methyl s-3-tert-butoxycarbonyl-2,2-dimethyl-4-oxazolidinecarboxylate, methyl s-n-boc-2,2-dimethyloxazolidine-4-carboxylate, methyl 4s-3-tert-butoxycarbonyl-2,2-dimethyloxazolidine-4-carboxylate, methyl r-n-boc-2,2-dimethyloxazolidine-4-carboxylate | |
| CC1(N(C(CO1)C(=O)OC)C(=O)OC(C)(C)C)C | |
| 259.302 | |
| 259.30 | |
| Liquid |
| 108149-60-6 | |
| C12H21NO5 | |
| 5 g | |
| ZNBUXTFASGDVCL-QMMMGPOBSA-N | |
| 3-O-tert-butyl 4-O-methyl (4S)-2,2-dimethyl-1,3-oxazolidine-3,4-dicarboxylate | |
| 688221 | |
| ≥96.0% (GC) |
Chemical Identifiers
| 108149-60-6 | |
| 259.302 | |
| ZNBUXTFASGDVCL-QMMMGPOBSA-N | |
| 688221 | |
| CC1(N(C(CO1)C(=O)OC)C(=O)OC(C)(C)C)C |
| C12H21NO5 | |
| MFCD00192279 | |
| s---3-tert-butoxycarbonyl-4-methoxycarbonyl-2,2-dimethyl-1,3-oxazolidine, 3-tert-butyl 4-methyl 4s-2,2-dimethyl-1,3-oxazolidine-3,4-dicarboxylate, methyl s---3-boc-2,2-dimethyl-4-oxazolidinecarboxylate, s-3-boc-2,2-dimethyl-oxazolidine-4-carboxylic acid methyl ester, s-3-tert-butyl 4-methyl 2,2-dimethyloxazolidine-3,4-dicarboxylate, methyl-s-3-tert-butoxycarbonyl-2,2-dimethyl-4-oxazolidinecarboxylate, methyl s-3-tert-butoxycarbonyl-2,2-dimethyl-4-oxazolidinecarboxylate, methyl s-n-boc-2,2-dimethyloxazolidine-4-carboxylate, methyl 4s-3-tert-butoxycarbonyl-2,2-dimethyloxazolidine-4-carboxylate, methyl r-n-boc-2,2-dimethyloxazolidine-4-carboxylate | |
| 3-O-tert-butyl 4-O-methyl (4S)-2,2-dimethyl-1,3-oxazolidine-3,4-dicarboxylate |
Safety and Handling
TSCA : No