missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(S)-3-Phenylcyclohexanone 98.0+%, TCI America™
Supplier: TCI America P15001G
Specifications
| (S)-3-Phenylcyclohexanone | |
| White | |
| MFCD08276411 | |
| CJAUDSQXFVZPTO-UHFFFAOYNA-N | |
| 3-phenylcyclohexan-1-one | |
| 11116555 | |
| ≥98.0% (GC) |
| 57344-86-2 | |
| C12H14O | |
| 1 g | |
| O=C1CCCC(C1)C1=CC=CC=C1 | |
| 174.24 | |
| 174.24 | |
| Crystalline Lumps |
Chemical Identifiers
| 57344-86-2 | |
| 174.24 | |
| CJAUDSQXFVZPTO-UHFFFAOYNA-N | |
| 3-phenylcyclohexan-1-one |
| C12H14O | |
| MFCD08276411 | |
| 11116555 | |
| O=C1CCCC(C1)C1=CC=CC=C1 |
Safety and Handling
TSCA : No
Recommended Storage : Refrigerator