missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(S)-3-Phenylcyclohexanone 98.0+%, TCI America™
$143.54 - $1427.87
Chemical Identifiers
| CAS | 57344-86-2 |
|---|---|
| Molecular Formula | C12H14O |
| Molecular Weight (g/mol) | 174.24 |
| MDL Number | MFCD08276411 |
| InChI Key | CJAUDSQXFVZPTO-UHFFFAOYNA-N |
| PubChem CID | 11116555 |
| IUPAC Name | 3-phenylcyclohexan-1-one |
| SMILES | O=C1CCCC(C1)C1=CC=CC=C1 |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
P1500100MG
|
TCI America
P1500100MG |
100 mg |
Each for $143.54
|
|
|||||
|
P15001G
|
TCI America
P15001G |
1 g |
Each for $1,427.87
|
|
|||||
Chemical Identifiers
| 57344-86-2 | |
| 174.24 | |
| CJAUDSQXFVZPTO-UHFFFAOYNA-N | |
| 3-phenylcyclohexan-1-one |
| C12H14O | |
| MFCD08276411 | |
| 11116555 | |
| O=C1CCCC(C1)C1=CC=CC=C1 |
Specifications
| 57344-86-2 | |
| C12H14O | |
| 100 mg | |
| O=C1CCCC(C1)C1=CC=CC=C1 | |
| 174.24 | |
| 174.24 | |
| Crystalline Lumps |
| White | |
| MFCD08276411 | |
| CJAUDSQXFVZPTO-UHFFFAOYNA-N | |
| 3-phenylcyclohexan-1-one | |
| 11116555 | |
| ≥98.0% (GC) | |
| (S)-3-Phenylcyclohexanone |
Safety and Handling
TSCA : No
Recommended Storage : Refrigerator