missing translation for 'onlineSavingsMsg'
Learn More

(S)-(+)-3-Methyl-2-butanol, 99%

Catalog No. p-7045106
Change view
Click to view available options
Quantity:
100 mg
500 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL09431MC 100 mg
AAL09431ME 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL09431MC Supplier Thermo Scientific Chemicals Supplier No. L09431MC
Only null left

CAS: 1517-66-4 | C5H12O | 88.15 g/mol

It is an pharmaceutical and organic intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is an pharmaceutical and organic intermediate.

Solubility
Soluble in alcohol. Insoluble in water.

Notes
Store in cool dry condition. Incompatible with strong oxidizing agents. Ensure ventilation. Protect from electrostatic charges.

Chemical Identifiers

CAS 1517-66-4
Molecular Formula C5H12O
Molecular Weight (g/mol) 88.15
MDL Number MFCD00064271
InChI Key MXLMTQWGSQIYOW-UHFFFAOYNA-N
Synonym s-+-3-methyl-2-butanol, 2s-3-methylbutan-2-ol, 2-butanol, 3-methyl-, 2s, +-3-methyl-2-butanol, 3-methyl-2-butanol #, s-3-methyl-2-butanol, s-3-methyl-butan-2-ol, unii-2nk7o363q6 component, unii-93ff0f303r component
PubChem CID 6999784
IUPAC Name (2S)-3-methylbutan-2-ol
SMILES CC(C)C(C)O

Specifications

Density 0.81
Boiling Point 112°C
Flash Point 26°C (78°F)
Refractive Index 1.409
Quantity 100 mg
UN Number UN1105
Beilstein 1718801
Solubility Information Soluble in alcohol. Insoluble in water.
Optical Rotation +5° (Neat)
Formula Weight 88.15
Percent Purity 99%
Chemical Name or Material (S)-(+)-3-Methyl-2-butanol
Show More Show Less
Hazard Category H226-H332-H335
Hazard Statement GHS H Statement
H226-H332-H335
Flammable liquid and vapor.
Harmful if inhaled.
May cause respiratory irritation.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P261-P271-P280-P303+P361+P353-P304+P340-P312-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: PENTANOLS
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only