CAS RN 1517-66-4
IUPAC Name:
3-methylbutan-2-ol
Synonyms:
3-methyl-2-butanol
Molecular Weight (g/mol):
88.15
Molecular Formula:
C5H12O
InChi Key:
MXLMTQWGSQIYOW-UHFFFAOYNA-N
SMILES:
CC(C)C(C)O
1
–
1
of
1
results
1
Promotions Available
(S)-(+)-3-Methyl-2-butanol, 99%
(S)-(+)-3-Methyl-2-butanol, 99%
CAS: 1517-66-4 Molecular Formula: C5H12O Molecular Weight (g/mol): 88.15 MDL Number: MFCD00064271 InChI Key: MXLMTQWGSQIYOW-UHFFFAOYNA-N Synonym: s-+-3-methyl-2-butanol,2s-3-methylbutan-2-ol,2-butanol, 3-methyl-, 2s,+-3-methyl-2-butanol,3-methyl-2-butanol #,s-3-methyl-2-butanol,s-3-methyl-butan-2-ol,unii-2nk7o363q6 component,unii-93ff0f303r component PubChem CID: 6999784 IUPAC Name: (2S)-3-methylbutan-2-ol SMILES: CC(C)C(C)O