Learn More
(S)-(-)-3-Hydroxypyrrolidine, 97+%, ee 98+%
CAS: 100243-39-8 | C4H10NO | 88.13 g/mol
Supplier: Thermo Scientific Chemicals L19498MD
Description
(S)-(-)-3-Hydroxypyrrolidine is an important raw material and intermediate used in organic synthesis, pharmaceuticals agrochemicals and dyestuff fields.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications(S)-(-)-3-Hydroxypyrrolidine is an important raw material and intermediate used in organic synthesis, pharmaceuticals agrochemicals and dyestuff fields.
Solubility
Fully miscible in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with oxidizing agents. It is hygroscopic and sensitive to air.
Specifications
| (S)-(-)-3-Hydroxypyrrolidine | |
| 15°C | |
| 108°C to 110°C (8 mmHg) | |
| ee 99% | |
| 1.49 | |
| 250 mg | |
| Air sensitive; Hygroscopic | |
| Fully miscible in water. | |
| −6.5° (c=3.5 in Methanol) | |
| (3S)-3-hydroxypyrrolidin-1-ium | |
| 2733874 | |
| 99% |
| 100243-39-8 | |
| 1.048 | |
| >110°C (230°F) | |
| C4H10NO | |
| MFCD00192426 | |
| 4652606 | |
| s-pyrrolidin-3-ol, s-3-hydroxypyrrolidine, 3s-pyrrolidin-3-ol, s-3-pyrrolidinol, s---3-pyrrolidinol, 3-pyrrolidinol, 3s, 3s-3-pyrrolidinol, 3s-3-hydroxypyrrolidine, s-3-hydroxy-pyrrolidine, s---3-hydroxypyrrolidine | |
| JHHZLHWJQPUNKB-BYPYZUCNSA-O | |
| O[C@H]1CC[NH2+]C1 | |
| 88.13 | |
| 87.12 |
Chemical Identifiers
| 100243-39-8 | |
| 88.13 | |
| JHHZLHWJQPUNKB-BYPYZUCNSA-O | |
| 2733874 |
| C4H10NO | |
| MFCD00192426 | |
| s-pyrrolidin-3-ol, s-3-hydroxypyrrolidine, 3s-pyrrolidin-3-ol, s-3-pyrrolidinol, s---3-pyrrolidinol, 3-pyrrolidinol, 3s, 3s-3-pyrrolidinol, 3s-3-hydroxypyrrolidine, s-3-hydroxy-pyrrolidine, s---3-hydroxypyrrolidine | |
| O[C@H]1CC[NH2+]C1 |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
TSCA : No
Recommended Storage : Ambient temperatures; Store under Argon
RUO – Research Use Only