missing translation for 'onlineSavingsMsg'
Learn More

(S)-(+)-3-Hydroxy-2,2-dimethylcyclohexanone 95.0+%, TCI America™

Catalog No. H0887100MG
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H0887100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. H0887100MG Supplier TCI America Supplier No. H0887100MG
Only null left

Chemical Identifiers

CAS 87655-21-8
Molecular Formula C8H14O2
Molecular Weight (g/mol) 142.198
MDL Number MFCD00671542
InChI Key MAGFJLNBFXFQGX-LURJTMIESA-N
PubChem CID 11018979
IUPAC Name (3S)-3-hydroxy-2,2-dimethylcyclohexan-1-one
SMILES CC1(C(CCCC1=O)O)C

Specifications

Color Yellow
Quantity 100 mg
Formula Weight 142.20
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material (S)-(+)-3-Hydroxy-2,2-dimethylcyclohexanone