accessibility menu, dialog, popup

UserName

H3COOHCH3

CAS RN 87655-21-8

IUPAC Name: 3-hydroxy-2,2-dimethylcyclohexan-1-one
Synonyms: 3-hydroxy-2,2-dimethylcyclohexan-1-one
Molecular Weight (g/mol): 142.2
Molecular Formula: C8H14O2
InChi Key: MAGFJLNBFXFQGX-UHFFFAOYNA-N
SMILES: CC1(C)C(O)CCCC1=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results