missing translation for 'onlineSavingsMsg'
Learn More

(S)-3-(Fmoc-amino)-3-(2-nitrophenyl)propionic acid, 95%

Catalog No. AAH5210103
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5210103 1 g
AAH5210106 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH5210103 Supplier Thermo Scientific Chemicals Supplier No. H5210103

CAS: 507472-25-5 | C24H20N2O6 | 432.43 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Solubility
Slightly soluble in water.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.

Chemical Identifiers

CAS 507472-25-5
Molecular Formula C24H20N2O6
Molecular Weight (g/mol) 432.43
MDL Number MFCD03428007
InChI Key DRESTDPDCGPKNI-UHFFFAOYNA-N
Synonym s-3-9h-fluoren-9-yl methoxy carbonyl amino-3-2-nitrophenyl propanoic acid, fmoc-s-3-amino-3-2-nitro-phenyl-propionic acid, fmoc-s-3-amino-3-2-nitrophenyl-propionic acid, fmoc-d-beta-phe 2-no2-oh, 3s-3-9h-fluoren-9-ylmethoxy carbonyl amino-3-2-nitrophenyl propanoic acid, pubchem24027, fmoc-?-phe 2-no2-oh, n-fmoc-2-nitro-d-beta-phenylalanine, fmoc-s-3-amino-3-2-nitrophenyl propionic acid
PubChem CID 7023399
IUPAC Name 3-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)-3-(2-nitrophenyl)propanoic acid
SMILES OC(=O)CC(NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12)C1=CC=CC=C1[N+]([O-])=O

Specifications

Melting Point 154°C to 156°C
Quantity 1 g
Solubility Information Slightly soluble in water.
Formula Weight 432.43
Percent Purity 95%
Chemical Name or Material (S)-3-(Fmoc-amino)-3-(2-nitrophenyl)propionic acid
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only