missing translation for 'onlineSavingsMsg'
Learn More

(S)-(-)-3-(BOC-Amino)pyrrolidine, 99%, Thermo Scientific™

$167.53 - $557.82

Chemical Identifiers

CAS 122536-76-9
Molecular Formula C9H18N2O2
Molecular Weight (g/mol) 186.26
InChI Key DQQJBEAXSOOCPG-ZETCQYMHSA-N
Synonym s-3-boc-amino pyrrolidine, s-tert-butyl pyrrolidin-3-ylcarbamate, s---3-boc-amino pyrrolidine, s-3-n-boc-aminopyrrolidine, tert-butyl n-3s-pyrrolidin-3-yl carbamate, s-+-3-boc-amino pyrrolidine, 3s---3-tert-butoxycarbonylamino pyrrolidine, tert-butyl s-pyrrolidin-3-ylcarbamate, s-3bocap, 3s---3-boc-amino pyrrolidine
PubChem CID 1514396
IUPAC Name tert-butyl N-[(3S)-pyrrolidin-3-yl]carbamate
SMILES CC(C)(C)OC(=O)NC1CCNC1
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
AC437900010
View Documents Promotion Details
N/A 1g
Each for $167.53
Only null left
 
AC437900050
View Documents Promotion Details
N/A 5g
Each for $557.82
Only null left
 
Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

122536-76-9
186.26
s-3-boc-amino pyrrolidine, s-tert-butyl pyrrolidin-3-ylcarbamate, s---3-boc-amino pyrrolidine, s-3-n-boc-aminopyrrolidine, tert-butyl n-3s-pyrrolidin-3-yl carbamate, s-+-3-boc-amino pyrrolidine, 3s---3-tert-butoxycarbonylamino pyrrolidine, tert-butyl s-pyrrolidin-3-ylcarbamate, s-3bocap, 3s---3-boc-amino pyrrolidine
tert-butyl N-[(3S)-pyrrolidin-3-yl]carbamate
C9H18N2O2
DQQJBEAXSOOCPG-ZETCQYMHSA-N
1514396
CC(C)(C)OC(=O)NC1CCNC1

Specifications

122536-76-9
100.0
98.5% min. (GC)
-20
DQQJBEAXSOOCPG-ZETCQYMHSA-N
tert-butyl N-[(3S)-pyrrolidin-3-yl]carbamate
1514396
99%
98.5
Authentic
C9H18N2O2
s-3-boc-amino pyrrolidine, s-tert-butyl pyrrolidin-3-ylcarbamate, s---3-boc-amino pyrrolidine, s-3-n-boc-aminopyrrolidine, tert-butyl n-3s-pyrrolidin-3-yl carbamate, s-+-3-boc-amino pyrrolidine, 3s---3-tert-butoxycarbonylamino pyrrolidine, tert-butyl s-pyrrolidin-3-ylcarbamate, s-3bocap, 3s---3-boc-amino pyrrolidine
CC(C)(C)OC(=O)NC1CCNC1
186.26
186.26
(S)-(-)-3-(BOC-amino)pyrrolidine
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes skin irritation.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously wit

GHS Signal Word: Warning

SDS
Documents

Documents

RUO – Research Use Only