missing translation for 'onlineSavingsMsg'
Learn More

(S)-(-)-3-(BOC-Amino)pyrrolidine, 99%

Catalog No. AC437900010
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC437900010 1 g
AC437900050 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC437900010 Supplier Thermo Scientific Chemicals Supplier No. 437900010
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 122536-76-9 | C9H18N2O2 | 186.26 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 122536-76-9
Molecular Formula C9H18N2O2
Molecular Weight (g/mol) 186.26
InChI Key DQQJBEAXSOOCPG-ZETCQYMHSA-N
Synonym s-3-boc-amino pyrrolidine, s-tert-butyl pyrrolidin-3-ylcarbamate, s---3-boc-amino pyrrolidine, s-3-n-boc-aminopyrrolidine, tert-butyl n-3s-pyrrolidin-3-yl carbamate, s-+-3-boc-amino pyrrolidine, 3s---3-tert-butoxycarbonylamino pyrrolidine, tert-butyl s-pyrrolidin-3-ylcarbamate, s-3bocap, 3s---3-boc-amino pyrrolidine
PubChem CID 1514396
IUPAC Name tert-butyl N-[(3S)-pyrrolidin-3-yl]carbamate
SMILES CC(C)(C)OC(=O)NC1CCNC1

Specifications

Melting Point 77.8°C to 80.7°C
CAS Min % 98.5
CAS Max % 100.0
Color White
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Quantity 1 g
Specific Rotation -20
Formula Weight 186.26
Percent Purity 99%
Physical Form Powder
Chemical Name or Material (S)-(-)-3-(BOC-amino)pyrrolidine
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes skin irritation.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously wit

RUO – Research Use Only