Learn More
(S)-(-)-3-(Boc-amino)pyrrolidine, 98%
CAS: 122536-76-9 | C9H18N2O2 | 186.255 g/mol
Chemical Identifiers
| CAS | 122536-76-9 |
|---|---|
| Molecular Formula | C9H18N2O2 |
| Molecular Weight (g/mol) | 186.255 |
| MDL Number | MFCD00143194 |
| InChI Key | DQQJBEAXSOOCPG-ZETCQYMHSA-N |
| Synonym | s-3-boc-amino pyrrolidine, s-tert-butyl pyrrolidin-3-ylcarbamate, s---3-boc-amino pyrrolidine, s-3-n-boc-aminopyrrolidine, tert-butyl n-3s-pyrrolidin-3-yl carbamate, s-+-3-boc-amino pyrrolidine, 3s---3-tert-butoxycarbonylamino pyrrolidine, tert-butyl s-pyrrolidin-3-ylcarbamate, s-3bocap, 3s---3-boc-amino pyrrolidine |
| PubChem CID | 1514396 |
| IUPAC Name | tert-butyl N-[(3S)-pyrrolidin-3-yl]carbamate |
| SMILES | CC(C)(C)OC(=O)NC1CCNC1 |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|
AAH5172906
|
N/A | 5 g |
N/A
|
|
|||||
|
AAH5172914
|
N/A | 25 g |
N/A
|
|
|||||
Description
(S)-(-)-3-(Boc-amino)pyrrolidine is used as pharmaceutical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications(S)-(-)-3-(Boc-amino)pyrrolidine is used as pharmaceutical intermediate.
Solubility
Soluble in water.
Notes
Air sensitive. Incompatible with oxidizing agents and carbon dioxide.
Chemical Identifiers
| 122536-76-9 | |
| 186.255 | |
| DQQJBEAXSOOCPG-ZETCQYMHSA-N | |
| 1514396 | |
| CC(C)(C)OC(=O)NC1CCNC1 |
| C9H18N2O2 | |
| MFCD00143194 | |
| s-3-boc-amino pyrrolidine, s-tert-butyl pyrrolidin-3-ylcarbamate, s---3-boc-amino pyrrolidine, s-3-n-boc-aminopyrrolidine, tert-butyl n-3s-pyrrolidin-3-yl carbamate, s-+-3-boc-amino pyrrolidine, 3s---3-tert-butoxycarbonylamino pyrrolidine, tert-butyl s-pyrrolidin-3-ylcarbamate, s-3bocap, 3s---3-boc-amino pyrrolidine | |
| tert-butyl N-[(3S)-pyrrolidin-3-yl]carbamate |
Specifications
| 50°C to 54°C | |
| C9H18N2O2 | |
| UN3259 | |
| s-3-boc-amino pyrrolidine, s-tert-butyl pyrrolidin-3-ylcarbamate, s---3-boc-amino pyrrolidine, s-3-n-boc-aminopyrrolidine, tert-butyl n-3s-pyrrolidin-3-yl carbamate, s-+-3-boc-amino pyrrolidine, 3s---3-tert-butoxycarbonylamino pyrrolidine, tert-butyl s-pyrrolidin-3-ylcarbamate, s-3bocap, 3s---3-boc-amino pyrrolidine | |
| DQQJBEAXSOOCPG-ZETCQYMHSA-N | |
| tert-butyl N-[(3S)-pyrrolidin-3-yl]carbamate | |
| 1514396 | |
| 186.25 | |
| 98% |
| 122536-76-9 | |
| MFCD00143194 | |
| 5377813 | |
| Soluble in water. | |
| CC(C)(C)OC(=O)NC1CCNC1 | |
| 186.255 | |
| −23° (c=1 in Methanol) | |
| Air Sensitive | |
| (S)-(-)-3-(Boc-amino)pyrrolidine |
Safety and Handling
GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
DOTInformation : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: AMINES, SOLID, CORROSIVE, N.O.S.
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only