missing translation for 'onlineSavingsMsg'
Learn More

(S)-3-Benzyloxycarbonylamino-2-(Boc-amino)propionic acid dicyclohexylammonium salt, 98%, Thermo Scientific Chemicals

Catalog No. p-7053843
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6155806 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH6155806 Supplier Thermo Scientific Chemicals Supplier No. H6155806

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 65710-58-9 | Molecular Formula: C28H45N3O6 | Molecular Weight: 519.68 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 65710-58-9
Molecular Formula C28H45N3O6
Molecular Weight (g/mol) 519.68
MDL Number MFCD00236880
InChI Key RPWGTQRQPVPFKR-UHFFFAOYNA-N
Synonym boc-dap z-oh dcha, dicyclohexylamine s-3-benzyloxy carbonyl amino-2-tert-butoxycarbonyl amino propanoate, boc-dap z-oh.dcha, boc-dap z-oh dicyclohexylammonium salt, pubchem23819, boc-dap z-oh?cha, boc-3-z-amino-l-alanine dicyclohexylammonium salt, boc-dap z-oh dicyclohexylammonium salt hplc, n, a-boc-n, a-z-l-2,3-diaminopropionic acid dicyclohexylammonium salt
PubChem CID 2755949
IUPAC Name N-cyclohexylcyclohexanamine;(2S)-2-[(2-methylpropan-2-yl)oxycarbonylamino]-3-(phenylmethoxycarbonylamino)propanoic acid
SMILES C1CCC(CC1)NC1CCCCC1.CC(C)(C)OC(=O)NC(CNC(=O)OCC1=CC=CC=C1)C(O)=O

Specifications

Beilstein 4100906
Quantity 5 g
Formula Weight 385.54
Percent Purity 98%
Physical Form Solid
Chemical Name or Material (S)-3-Benzyloxycarbonylamino-2-(Boc-amino)propionic acid dicyclohexylammonium salt
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only