accessibility menu, dialog, popup

UserName

CAS RN 65710-58-9

IUPAC Name: 3-{[(benzyloxy)carbonyl]amino}-2-{[(tert-butoxy)carbonyl]amino}propanoic acid; N-cyclohexylcyclohexanamine
Synonyms: 3-{[(benzyloxy)carbonyl]amino}-2-[(tert-butoxycarbonyl)amino]propanoic acid; dicha
Molecular Weight (g/mol): 519.68
Molecular Formula: C28H45N3O6
InChi Key: RPWGTQRQPVPFKR-UHFFFAOYNA-N
SMILES: C1CCC(CC1)NC1CCCCC1.CC(C)(C)OC(=O)NC(CNC(=O)OCC1=CC=CC=C1)C(O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)