missing translation for 'onlineSavingsMsg'
Learn More

(S)-3-Amino-5-hexynoic acid hydrochloride, 95%

Catalog No. AAH5201003
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5201003 1 g
AAH52010MD 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH5201003 Supplier Thermo Scientific Chemicals Supplier No. H5201003
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 270596-46-8 | C6H10ClNO2 | 163.601 g/mol

(S)-3-Amino-5-hexynoic acid hydrochloride is used for labeling and detection purpose.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
(S)-3-Amino-5-hexynoic acid hydrochloride is used for labeling and detection purpose.

Solubility
Soluble in water (90 g/L at 25°C).

Notes
Air sensitive. Incompatible with oxidizing agents.

Chemical Identifiers

CAS 270596-46-8
Molecular Formula C6H10ClNO2
Molecular Weight (g/mol) 163.601
MDL Number MFCD01861006
InChI Key FVKOZZHCHSRKJA-JEDNCBNOSA-N
Synonym s-3-amino-5-hexynoic acid hydrochloride, s-3-aminohex-5-ynoic acid hydrochloride, 3s-3-aminohex-5-ynoic acid hydrochloride, s-3-amino-5-hexynoic acid-hcl, l-beta-homopropargylglycine hydrochloride, 5-hexynoic acid, 3-amino-, 3s-, hydrochloride 1:1, s-3-amino-5-hexynoic acidhcl, l-ss-homopropargylglycine hydrochloride
PubChem CID 2761700
IUPAC Name (3S)-3-aminohex-5-ynoic acid;hydrochloride
SMILES C#CCC(CC(=O)O)N.Cl

Specifications

Solubility Information Soluble in water (90g/L at 25°C).
Quantity 1 g
Formula Weight 163.6
Sensitivity Air Sensitive
Percent Purity 95%
Chemical Name or Material (S)-3-Amino-5-hexynoic acid hydrochloride
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures; Store under Argon

RUO – Research Use Only