accessibility menu, dialog, popup

UserName

HCOOHNH2HCl

CAS RN 270596-46-8

IUPAC Name: 3-aminohex-5-ynoic acid hydrochloride
Synonyms: 3-aminohex-5-ynoic acid hydrochloride
Molecular Weight (g/mol): 163.6
Molecular Formula: C6H10ClNO2
InChi Key: FVKOZZHCHSRKJA-UHFFFAOYNA-N
SMILES: Cl.NC(CC#C)CC(O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (1)
  • (1)