missing translation for 'onlineSavingsMsg'
Learn More

(S)-3-Amino-3-(3-nitrophenyl)propionic acid, 95%

Catalog No. AAH5202706
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5202706 5 g
AAH5202703 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH5202706 Supplier Thermo Scientific Chemicals Supplier No. H5202706
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 734529-57-8 | C9H10N2O4 | 210.189 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 734529-57-8
Molecular Formula C9H10N2O4
Molecular Weight (g/mol) 210.189
MDL Number MFCD04113693
InChI Key SJBFILRQMRECCK-QMMMGPOBSA-N
Synonym s-3-amino-3-3-nitrophenyl propionic acid, s-3-amino-3-3-nitrophenyl propanoic acid, s-beta-3-nitrophenyl alanine, s-3-3-nitrophenyl-beta-alanine, 3s-3-amino-3-3-nitrophenyl propanoic acid, h-beta-phe 3-no2-oh, s-3-amino-3-3-nitro-phenyl-propionic acid, pubchem15094, 3-nitro-l-beta-phenylalanine
PubChem CID 706689
SMILES C1=CC(=CC(=C1)[N+](=O)[O-])C(CC(=O)O)N

Specifications

Quantity 5 g
Formula Weight 210.19
Sensitivity Air Sensitive
Percent Purity 95%
Chemical Name or Material (S)-3-Amino-3-(3-nitrophenyl)propionic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only