accessibility menu, dialog, popup

UserName

NOOOHOH2N

CAS RN 734529-57-8

IUPAC Name: 3-amino-3-(3-nitrophenyl)propanoic acid
Synonyms: 3-amino-3-(3-nitrophenyl)propanoic acid
Molecular Weight (g/mol): 210.19
Molecular Formula: C9H10N2O4
InChi Key: SJBFILRQMRECCK-UHFFFAOYNA-N
SMILES: NC(CC(O)=O)C1=CC(=CC=C1)[N+]([O-])=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (1)
  • (1)