missing translation for 'onlineSavingsMsg'
Learn More

(S)-3-Amino-2-(carbobenzoxyamino)propionic Acid 98.0+%, TCI America™

Catalog No. p-7117929
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A24711G 1 g
A24715G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A24711G Supplier TCI America Supplier No. A24711G
Only null left

Chemical Identifiers

CAS 35761-26-3
Molecular Formula C11H14N2O4
Molecular Weight (g/mol) 238.243
MDL Number MFCD00237345
InChI Key FOXRXVSTFGNURG-VIFPVBQESA-N
Synonym z-dap-oh, cbz-beta-amino-l-alanine, 3-amino-2-n-cbz-l-alanine, s-3-amino-2-benzyloxy carbonyl amino propanoic acid, cbz-l-dap-oh, s-3-amino-2-benzyloxycarbonylamino propanoic acid, cbz-dap-oh, h-dap z-oh, n-alpha-cbz-l-2,3-diaminopropionic acid, z-dpr-oh
PubChem CID 2756313
IUPAC Name (2S)-3-amino-2-(phenylmethoxycarbonylamino)propanoic acid
SMILES C1=CC=C(C=C1)COC(=O)NC(CN)C(=O)O

Specifications

Melting Point 230°C
Color White
Quantity 1 g
Formula Weight 238.24
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material (S)-3-Amino-2-(carbobenzoxyamino)propionic Acid