accessibility menu, dialog, popup

UserName

CAS RN 35761-26-3

IUPAC Name: 3-amino-2-{[(benzyloxy)carbonyl]amino}propanoic acid
Synonyms: 3-amino-2-{[(benzyloxy)carbonyl]amino}propanoic acid
Molecular Weight (g/mol): 238.24
Molecular Formula: C11H14N2O4
InChi Key: FOXRXVSTFGNURG-UHFFFAOYNA-N
SMILES: NCC(NC(=O)OCC1=CC=CC=C1)C(O)=O
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)