missing translation for 'onlineSavingsMsg'
Learn More

(S)-(+)-3-Amino-1-Boc-piperidine, 97%

CAS: 625471-18-3 | C10H20N2O2 | 200.28 g/mol

$381.30 - $1265.73

Chemical Identifiers

CAS 625471-18-3
Molecular Formula C10H20N2O2
Molecular Weight (g/mol) 200.28
MDL Number MFCD03094718
InChI Key AKQXKEBCONUWCL-UHFFFAOYNA-N
Synonym s-1-boc-3-aminopiperidine, s-3-amino-1-n-boc-piperidine, tert-butyl 3s-3-aminopiperidine-1-carboxylate, s-tert-butyl 3-aminopiperidine-1-carboxylate, s-3-amino-1-boc-piperidine, s-+-3-amino-1-boc-piperidine, 3s-3-aminopiperidine, n1-boc protected, s-1-boc-3-amniopiperidine, s-3-amino-piperidine-1-carboxylic acid tert-butyl ester, s-3-amino-n-tboc-piperidine
PubChem CID 1501975
IUPAC Name tert-butyl (3S)-3-aminopiperidine-1-carboxylate
SMILES CC(C)(C)OC(=O)N1CCCC(N)C1
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH3053803
View Documents
Thermo Scientific Chemicals
H3053803
1 g
Each for $381.30
Only null left
 
AAH3053806
View Documents
Thermo Scientific Chemicals
H3053806
5 g
Each for $1,265.73
Only null left
 
Description

Description

(S)-(+)-3-Amino-1-Boc-piperidine is used to prepare selective noncovalent inhibitors of the bacterial cysteine protease IdeS. The product obtained by the reductive elimination reaction with ethylglyoxalte is used as a reference compound, which is used to determine the absolute configuration of the two enantiomers.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
(S)-(+)-3-Amino-1-Boc-piperidine is used to prepare selective noncovalent inhibitors of the bacterial cysteine protease IdeS. The product obtained by the reductive elimination reaction with ethylglyoxalte is used as a reference compound, which is used to determine the absolute configuration of the two enantiomers.

Solubility
Soluble in dimethylsulfoxide.

Notes
Air and carbon dioxide sensitive. Incompatible with strong oxidizing agents, strong acids and bases. Store in a cool place.
Specifications

Chemical Identifiers

625471-18-3
200.28
AKQXKEBCONUWCL-UHFFFAOYNA-N
1501975
CC(C)(C)OC(=O)N1CCCC(N)C1
C10H20N2O2
MFCD03094718
s-1-boc-3-aminopiperidine, s-3-amino-1-n-boc-piperidine, tert-butyl 3s-3-aminopiperidine-1-carboxylate, s-tert-butyl 3-aminopiperidine-1-carboxylate, s-3-amino-1-boc-piperidine, s-+-3-amino-1-boc-piperidine, 3s-3-aminopiperidine, n1-boc protected, s-1-boc-3-amniopiperidine, s-3-amino-piperidine-1-carboxylic acid tert-butyl ester, s-3-amino-n-tboc-piperidine
tert-butyl (3S)-3-aminopiperidine-1-carboxylate

Specifications

625471-18-3
MFCD03094718
UN2735
s-1-boc-3-aminopiperidine, s-3-amino-1-n-boc-piperidine, tert-butyl 3s-3-aminopiperidine-1-carboxylate, s-tert-butyl 3-aminopiperidine-1-carboxylate, s-3-amino-1-boc-piperidine, s-+-3-amino-1-boc-piperidine, 3s-3-aminopiperidine, n1-boc protected, s-1-boc-3-amniopiperidine, s-3-amino-piperidine-1-carboxylic acid tert-butyl ester, s-3-amino-n-tboc-piperidine
AKQXKEBCONUWCL-UHFFFAOYNA-N
tert-butyl (3S)-3-aminopiperidine-1-carboxylate
1501975
97%
C10H20N2O2
1 g
Air Sensitive
Soluble in dimethylsulfoxide.
CC(C)(C)OC(=O)N1CCCC(N)C1
200.28
200.28
(S)-(+)-3-Amino-1-Boc-piperidine
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.

P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c

H314-H335

DOTInformation : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: AMINES, LIQUID, CORROSIVE, N.O.S.

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

RUO – Research Use Only